Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HAPLN4
Entrez Gene ID
404037
Gene Name
hyaluronan and proteoglycan link protein 4
Gene Aliases
BRAL2
Chromosome Location
Chr.19: 19255641 - 19262787 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023002.2 NP_075378.1 2 - 3 173 68
GenBank mRNA AB107883.1 - 1 - 2 118 68
AY262756.1 - 2 - 3 137 68
BC142644.1 - 2 - 3 173 68
BC142698.1 - 2 - 3 182 68

View Full Product Details