The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023002.2 | NP_075378.1 | 3 - 4 | 540 | 83 |
GenBank mRNA | AB107883.1 | - | 2 - 3 | 485 | 83 |
AY262756.1 | - | 3 - 4 | 504 | 83 | |
BC142644.1 | - | 3 - 4 | 540 | 83 | |
BC142698.1 | - | 3 - 4 | 549 | 83 |