The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175736.4 | NP_783863.4 | 2 - 3 | 451 | 146 |
NM_198900.2 | NP_944489.2 | 2 - 3 | 451 | 146 | |
XM_005269218.2 | XP_005269275.1 | 2 - 3 | 430 | 146 | |
XM_011538968.2 | XP_011537270.1 | 2 - 3 | 920 | 146 | |
XM_011538969.2 | XP_011537271.1 | 2 - 3 | 919 | 146 | |
XM_011538970.2 | XP_011537272.1 | 2 - 3 | 922 | 146 | |
XM_011538971.2 | XP_011537273.1 | 2 - 3 | 495 | 146 | |
XM_011538972.2 | XP_011537274.1 | 2 - 3 | 918 | 146 | |
XM_011538974.2 | XP_011537276.1 | 3 - 4 | 252 | 146 | |
GenBank mRNA | AB095934.2 | - | 2 - 3 | 451 | 146 |
AK128195.1 | - | 2 - 3 | 431 | 146 | |
BC159100.1 | - | 2 - 3 | 451 | 146 | |
BQ707902.1 | - | 2 - 3 | 112 | 146 |