The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175736.4 | NP_783863.4 | 21 - 22 | 2752 | 61 |
NM_198900.2 | NP_944489.2 | 20 - 21 | 2599 | 61 | |
XM_005269218.2 | XP_005269275.1 | 21 - 22 | 2731 | 61 | |
XM_011538968.2 | XP_011537270.1 | 21 - 22 | 3221 | 61 | |
XM_011538969.2 | XP_011537271.1 | 21 - 22 | 3220 | 61 | |
XM_011538970.2 | XP_011537272.1 | 21 - 22 | 3220 | 61 | |
XM_011538971.2 | XP_011537273.1 | 21 - 22 | 2796 | 61 | |
XM_011538972.2 | XP_011537274.1 | 20 - 21 | 3066 | 61 | |
XM_011538973.2 | XP_011537275.2 | 17 - 18 | 2211 | 61 | |
XM_011538974.2 | XP_011537276.1 | 22 - 23 | 2553 | 61 | |
GenBank mRNA | AB095934.2 | - | 21 - 22 | 2752 | 61 |
AK127200.1 | - | 8 - 9 | 1225 | 61 | |
AK128195.1 | - | 20 - 21 | 2579 | 61 | |
AL162062.1 | - | 5 - 6 | 709 | 61 | |
BC033181.1 | - | 5 - 6 | 536 | 61 | |
BC159100.1 | - | 21 - 22 | 2752 | 61 |