Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RASSF4
Entrez Gene ID
83937
Gene Name
Ras association domain family member 4
Gene Aliases
AD037
Chromosome Location
Chr.10: 44959753 - 44994724 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032023.3 NP_114412.2 9 - 10 943 66
XM_005271833.4 XP_005271890.1 9 - 10 993 66
XM_005271835.4 XP_005271892.1 9 - 10 943 66
XM_006718013.3 XP_006718076.1 9 - 10 1038 66
XM_006718014.3 XP_006718077.1 8 - 9 2457 66
XM_011540255.2 XP_011538557.1 8 - 9 818 66
XM_017016754.1 XP_016872243.1 8 - 9 2583 66
XM_017016755.1 XP_016872244.1 8 - 9 2538 66
XM_017016756.1 XP_016872245.1 9 - 10 1064 66
XM_017016757.1 XP_016872246.1 8 - 9 2609 66
XM_017016758.1 XP_016872247.1 7 - 8 2314 66
GenBank mRNA AB209446.1 - 8 - 9 2815 66
AF260335.1 - 9 - 10 907 66
AK055763.1 - 7 - 8 1951 66
AK092770.1 - 6 - 7 2154 66
AK097272.1 - 4 - 5 2221 66
AL713716.1 - 1 - 2 116 66
AY216713.1 - 9 - 10 943 66
AY216714.1 - 9 - 10 903 66
AY216715.1 - 9 - 10 970 66
AY216716.1 - 8 - 9 851 66
BC032593.1 - 9 - 10 925 66

View Full Product Details