The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178160.2 | NP_835454.1 | 6 - 7 | 1616 | 126 |
XM_011525479.1 | XP_011523781.1 | 7 - 8 | 1826 | 126 |