Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CXCL6
Entrez Gene ID
6372
Gene Name
C-X-C motif chemokine ligand 6
Gene Aliases
CKA-3, GCP-2, GCP2, SCYB6
Chromosome Location
Chr.4: 73836556 - 73838760 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002993.3 NP_002984.1 2 - 3 441 125
GenBank mRNA AK311981.1 - 2 - 3 318 125
BC013744.1 - 2 - 3 276 125
BP204144.1 - 2 - 3 441 125
U81234.1 - 2 - 3 309 125
Y08770.1 - 1 - 2 135 125

View Full Product Details