The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004988.4 | NP_004979.3 | 1 - 2 | 85 | 124 |
GenBank mRNA | AK314979.1 | - | 1 - 2 | 85 | 124 |
AY148486.1 | - | 1 - 2 | 43 | 124 | |
BC017555.1 | - | 1 - 2 | 52 | 124 | |
DB053667.1 | - | 1 - 2 | 99 | 124 | |
EU161102.1 | - | 1 - 2 | 43 | 124 |