The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005313.4 | NP_005304.3 | 1 - 2 | 316 | 129 |
GenBank mRNA | BC014433.1 | - | 1 - 2 | 257 | 129 |
D83485.1 | - | 1 - 2 | 190 | 129 |