Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40393936-40394199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRX
Entrez Gene ID
57716
Gene Name
periaxin
Gene Aliases
CMT4F
UniGene
Hs.205457
Gene Chromosome Location
Chr.19: 40393762-40414718 on Build GRCh38
Transcripts
NM_020956.2 NM_181882.2 XM_011527171.2 XM_017027046.1 XM_017027047.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1663714 c.4356G>C p.R1452S Chr.19: 40393996-40393996 on Build GRCh38
4581041 c.4186G>A p.D1396N Chr.19: 40394166-40394166 on Build GRCh38
5855784 c.4202C>T p.S1401F Chr.19: 40394150-40394150 on Build GRCh38
4140645 c.4270G>T p.G1424W Chr.19: 40394082-40394082 on Build GRCh38
4547817 c.4321G>T p.G1441W Chr.19: 40394031-40394031 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757627373 Chr.19: 40394017-40394017 on Build GRCh38
rs368067072 Chr.19: 40394019-40394019 on Build GRCh38
rs369949312 Chr.19: 40394003-40394003 on Build GRCh38
rs752857860 Chr.19: 40393993-40393993 on Build GRCh38
rs146468976 Chr.19: 40394133-40394133 on Build GRCh38
rs780827123 Chr.19: 40394033-40394033 on Build GRCh38
rs199974998 Chr.19: 40394068-40394068 on Build GRCh38
rs770666481 Chr.19: 40394040-40394040 on Build GRCh38
rs368827070 Chr.19: 40394045-40394045 on Build GRCh38
rs760540823 Chr.19: 40394152-40394152 on Build GRCh38
rs201222650 Chr.19: 40394109-40394109 on Build GRCh38
rs878855285 Chr.19: 40394048-40394048 on Build GRCh38
rs891393791 Chr.19: 40394091-40394091 on Build GRCh38
rs377083046 Chr.19: 40394038-40394038 on Build GRCh38
rs777172895 Chr.19: 40394101-40394101 on Build GRCh38
rs751443099 Chr.19: 40393977-40393977 on Build GRCh38
rs767543289 Chr.19: 40393988-40393988 on Build GRCh38
rs866708515 Chr.19: 40394075-40394075 on Build GRCh38
rs199895355 Chr.19: 40394039-40394039 on Build GRCh38
rs964490425 Chr.19: 40394122-40394122 on Build GRCh38
rs778270475 Chr.19: 40394016-40394017 on Build GRCh38
rs765667981 Chr.19: 40394088-40394088 on Build GRCh38
rs1003836485 Chr.19: 40394057-40394057 on Build GRCh38
rs375802521 Chr.19: 40394099-40394099 on Build GRCh38
rs771992595 Chr.19: 40394146-40394146 on Build GRCh38
rs758960812 Chr.19: 40394090-40394090 on Build GRCh38
rs373051724 Chr.19: 40393976-40393976 on Build GRCh38
rs771234852 Chr.19: 40394105-40394106 on Build GRCh38
rs983272464 Chr.19: 40394179-40394179 on Build GRCh38
rs779727296 Chr.19: 40394135-40394136 on Build GRCh38
rs764183288 Chr.19: 40394156-40394156 on Build GRCh38
rs1005254872 Chr.19: 40394120-40394120 on Build GRCh38
rs747327914 Chr.19: 40394017-40394018 on Build GRCh38
rs754032938 Chr.19: 40394082-40394082 on Build GRCh38
rs780680917 Chr.19: 40394094-40394094 on Build GRCh38
rs759435622 Chr.19: 40394051-40394051 on Build GRCh38
rs776616217 Chr.19: 40394164-40394164 on Build GRCh38
rs746338096 Chr.19: 40394018-40394018 on Build GRCh38
rs1056637272 Chr.19: 40394070-40394070 on Build GRCh38
rs372280596 Chr.19: 40394035-40394035 on Build GRCh38
rs143125201 Chr.19: 40394009-40394009 on Build GRCh38
rs267605484 Chr.19: 40394104-40394104 on Build GRCh38
rs758967287 Chr.19: 40394013-40394015 on Build GRCh38
rs142436702 Chr.19: 40394087-40394087 on Build GRCh38
rs917161355 Chr.19: 40394180-40394180 on Build GRCh38
rs139051512 Chr.19: 40394145-40394145 on Build GRCh38
rs775415523 Chr.19: 40394060-40394060 on Build GRCh38
rs138437458 Chr.19: 40394034-40394034 on Build GRCh38
rs775431247 Chr.19: 40394150-40394150 on Build GRCh38
rs566745894 Chr.19: 40394166-40394166 on Build GRCh38
rs149093940 Chr.19: 40393986-40393986 on Build GRCh38
rs866036088 Chr.19: 40394097-40394097 on Build GRCh38
rs895345246 Chr.19: 40394085-40394085 on Build GRCh38
rs747480005 Chr.19: 40394095-40394095 on Build GRCh38
rs911683158 Chr.19: 40394028-40394028 on Build GRCh38
rs533966999 Chr.19: 40394121-40394121 on Build GRCh38
rs377062046 Chr.19: 40393975-40393975 on Build GRCh38
rs760875742 Chr.19: 40394061-40394061 on Build GRCh38
rs767488196 Chr.19: 40394052-40394052 on Build GRCh38
rs750814264 Chr.19: 40394178-40394178 on Build GRCh38
rs768051108 Chr.19: 40394002-40394002 on Build GRCh38

View Full Product Details