The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_002734.2 | 1 - 1 | 192 | 154 | |
GenBank mRNA | AF095289.1 | - | 1 - 1 | 163 | 154 |