The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277.2 | NP_001268.2 | 5 - 6 | 978 | 158 |
NM_212469.1 | NP_997634.1 | 4 - 5 | 924 | 158 | |
XM_017017147.1 | XP_016872636.1 | 4 - 5 | 862 | 158 | |
XM_017017148.1 | XP_016872637.1 | 4 - 5 | 2334 | 158 | |
GenBank mRNA | BC036471.2 | - | 4 - 5 | 802 | 158 |
D10704.1 | - | 5 - 6 | 788 | 158 |