Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GOLGA1
Entrez Gene ID
2800
Gene Name
golgin A1
Gene Aliases
golgin-97
Chromosome Location
Chr.9: 124878294 - 124941114 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002077.3 NP_002068.1 16 - 17 1839 66
XM_005251929.3 XP_005251986.1 16 - 17 1771 66
XM_006717062.3 XP_006717125.1 16 - 17 1835 66
XM_006717063.3 XP_006717126.1 16 - 17 1806 66
GenBank mRNA AK302445.1 - 16 - 17 1648 66
BC032853.1 - 16 - 17 1809 66
U51587.1 - 16 - 17 1831 66

View Full Product Details