The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004687.4 | NP_004678.3 | 17 - 18 | 3432 | 68 |
XM_005257784.2 | XP_005257841.1 | 16 - 17 | 3581 | 68 | |
XM_005257785.4 | XP_005257842.1 | 18 - 19 | 3549 | 68 | |
XM_005257786.4 | XP_005257843.1 | 17 - 18 | 4336 | 68 | |
XM_006722168.3 | XP_006722231.1 | 17 - 18 | 3376 | 68 | |
XM_011525460.2 | XP_011523762.1 | 18 - 19 | 4429 | 68 | |
GenBank mRNA | AB014547.2 | - | 15 - 16 | 3322 | 68 |
AF264717.1 | - | 17 - 18 | 3375 | 68 | |
BC035609.1 | - | 17 - 18 | 3393 | 68 |