The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034194.1 | NP_001029366.1 | 6 - 7 | 711 | 73 |
NM_005033.2 | NP_005024.2 | 6 - 7 | 711 | 73 | |
XM_011532035.2 | XP_011530337.1 | 6 - 7 | 733 | 73 | |
GenBank mRNA | AJ505989.1 | - | 6 - 7 | 708 | 73 |
AJ517294.1 | - | 6 - 7 | 708 | 73 | |
AK301967.1 | - | 6 - 7 | 710 | 73 | |
AK307548.1 | - | 6 - 7 | 650 | 73 | |
AK310894.1 | - | 2 - 3 | 107 | 73 | |
BX649106.1 | - | 6 - 7 | 745 | 73 | |
M58460.1 | - | 5 - 6 | 770 | 73 | |
U09215.1 | - | 4 - 5 | 351 | 73 |