The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034194.1 | NP_001029366.1 | 7 - 8 | 844 | 71 |
NM_005033.2 | NP_005024.2 | 7 - 8 | 844 | 71 | |
XM_011532035.2 | XP_011530337.1 | 7 - 8 | 866 | 71 | |
GenBank mRNA | AJ505989.1 | - | 7 - 8 | 841 | 71 |
AJ517294.1 | - | 7 - 8 | 841 | 71 | |
AK301967.1 | - | 7 - 8 | 843 | 71 | |
AK307548.1 | - | 7 - 8 | 783 | 71 | |
AK310894.1 | - | 3 - 4 | 240 | 71 | |
BC142978.1 | - | 7 - 8 | 815 | 71 | |
M58460.1 | - | 6 - 7 | 903 | 71 | |
U09215.1 | - | 5 - 6 | 484 | 71 |