Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POLR2H
Entrez Gene ID
5437
Gene Name
RNA polymerase II subunit H
Gene Aliases
RPABC3, RPB17, RPB8
Chromosome Location
Chr.3: 184361710 - 184368595 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278699.1 NP_001265628.1 4 - 5 855 118
NM_001278700.1 NP_001265629.1 4 - 5 696 118
NM_006232.3 NP_006223.2 4 - 5 846 118
XM_006713668.3 XP_006713731.1 5 - 6 1393 118
XM_006713670.2 XP_006713733.1 5 - 6 1393 118
GenBank mRNA BC000739.2 - 4 - 5 396 118
BM541682.1 - 4 - 5 653 118
U37689.1 - 4 - 5 501 118
Z49199.1 - 4 - 5 414 118

View Full Product Details