Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST18
Entrez Gene ID
9705
Gene Name
ST18, C2H2C-type zinc finger
Gene Aliases
NZF3, ZC2H2C3, ZC2HC10, ZNF387
Chromosome Location
Chr.8: 52110832 - 52409879 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014682.2 NP_055497.1 10 - 11 1891 70
XM_006716487.1 XP_006716550.1 10 - 11 1658 70
XM_011517629.1 XP_011515931.1 11 - 12 1814 70
XM_011517631.1 XP_011515933.1 12 - 13 1855 70
XM_011517632.1 XP_011515934.1 10 - 11 1669 70
XM_011517633.1 XP_011515935.1 10 - 11 1662 70
XM_011517634.1 XP_011515936.1 10 - 11 1858 70
XM_011517635.1 XP_011515937.1 10 - 11 1862 70
XM_011517636.2 XP_011515938.1 9 - 10 1939 70
XM_011517637.1 XP_011515939.1 8 - 9 1396 70
XM_011517638.2 XP_011515940.1 7 - 8 1437 70
XM_011517641.1 XP_011515943.1 12 - 13 1902 70
XM_011517642.1 XP_011515944.1 11 - 12 1814 70
XM_017014047.1 XP_016869536.1 10 - 11 2018 70
XM_017014048.1 XP_016869537.1 10 - 11 2022 70
XM_017014049.1 XP_016869538.1 11 - 12 2068 70
XM_017014050.1 XP_016869539.1 10 - 11 1855 70
XM_017014051.1 XP_016869540.1 9 - 10 1862 70
XM_017014052.1 XP_016869541.1 11 - 12 2072 70
XM_017014053.1 XP_016869542.1 11 - 12 2059 70
XM_017014054.1 XP_016869543.1 9 - 10 2108 70
XM_017014055.1 XP_016869544.1 9 - 10 1699 70
XM_017014056.1 XP_016869545.1 12 - 13 2109 70
XM_017014057.1 XP_016869546.1 10 - 11 1912 70
XM_017014058.1 XP_016869547.1 10 - 11 1673 70
XM_017014059.1 XP_016869548.1 10 - 11 1859 70
XM_017014060.1 XP_016869549.1 11 - 12 1896 70
XM_017014061.1 XP_016869550.1 9 - 10 2031 70
XM_017014062.1 XP_016869551.1 8 - 9 1783 70
XM_017014063.1 XP_016869552.1 9 - 10 2329 70
XM_017014064.1 XP_016869553.1 9 - 10 1943 70
XM_017014065.1 XP_016869554.1 11 - 12 2063 70
XM_017014066.1 XP_016869555.1 9 - 10 1702 70
XM_017014067.1 XP_016869556.1 9 - 10 2112 70
XM_017014068.1 XP_016869557.1 10 - 11 1903 70
XM_017014069.1 XP_016869558.1 7 - 8 1600 70
XM_017014070.1 XP_016869559.1 6 - 7 1521 70
XM_017014071.1 XP_016869560.1 7 - 8 1769 70
XM_017014072.1 XP_016869561.1 11 - 12 1933 70
XM_017014073.1 XP_016869562.1 11 - 12 1571 70
XM_017014074.1 XP_016869563.1 10 - 11 1854 70
XM_017014075.1 XP_016869564.1 11 - 12 2105 70
XM_017014076.1 XP_016869565.1 11 - 12 2109 70
XM_017014077.1 XP_016869566.1 11 - 12 1746 70
XM_017014078.1 XP_016869567.1 10 - 11 2026 70
XM_017014079.1 XP_016869568.1 10 - 11 2417 70
XM_017014080.1 XP_016869569.1 8 - 9 1658 70
XM_017014081.1 XP_016869570.1 10 - 11 2025 70
XM_017014082.1 XP_016869571.1 10 - 11 1860 70
XM_017014083.1 XP_016869572.1 9 - 10 1700 70
XM_017014084.1 XP_016869573.1 7 - 8 1438 70
GenBank mRNA AB011107.1 - 10 - 11 1755 70
BC031075.1 - 10 - 11 1788 70
BC117147.1 - 5 - 6 1148 70
BC117149.1 - 5 - 6 1148 70
BC143872.1 - 5 - 6 1143 70
BC143873.1 - 6 - 7 1236 70

View Full Product Details