Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST18
Entrez Gene ID
9705
Gene Name
ST18, C2H2C-type zinc finger
Gene Aliases
NZF3, ZC2H2C3, ZC2HC10, ZNF387
Chromosome Location
Chr.8: 52110832 - 52409879 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014682.2 NP_055497.1 22 - 23 3486 69
XM_006716487.1 XP_006716550.1 22 - 23 3253 69
XM_011517629.1 XP_011515931.1 23 - 24 3409 69
XM_011517631.1 XP_011515933.1 24 - 25 3450 69
XM_011517632.1 XP_011515934.1 22 - 23 3264 69
XM_011517633.1 XP_011515935.1 22 - 23 3257 69
XM_011517634.1 XP_011515936.1 22 - 23 3453 69
XM_011517635.1 XP_011515937.1 22 - 23 3457 69
XM_011517636.2 XP_011515938.1 21 - 22 3534 69
XM_011517637.1 XP_011515939.1 20 - 21 2991 69
XM_011517638.2 XP_011515940.1 19 - 20 3032 69
XM_011517641.1 XP_011515943.1 24 - 25 3497 69
XM_011517642.1 XP_011515944.1 22 - 23 3163 69
XM_017014047.1 XP_016869536.1 22 - 23 3613 69
XM_017014048.1 XP_016869537.1 22 - 23 3617 69
XM_017014049.1 XP_016869538.1 23 - 24 3663 69
XM_017014050.1 XP_016869539.1 22 - 23 3450 69
XM_017014051.1 XP_016869540.1 21 - 22 3457 69
XM_017014052.1 XP_016869541.1 23 - 24 3667 69
XM_017014053.1 XP_016869542.1 23 - 24 3654 69
XM_017014054.1 XP_016869543.1 21 - 22 3703 69
XM_017014055.1 XP_016869544.1 21 - 22 3294 69
XM_017014056.1 XP_016869545.1 24 - 25 3704 69
XM_017014057.1 XP_016869546.1 22 - 23 3507 69
XM_017014058.1 XP_016869547.1 22 - 23 3268 69
XM_017014059.1 XP_016869548.1 22 - 23 3454 69
XM_017014060.1 XP_016869549.1 23 - 24 3491 69
XM_017014061.1 XP_016869550.1 21 - 22 3626 69
XM_017014062.1 XP_016869551.1 20 - 21 3378 69
XM_017014063.1 XP_016869552.1 21 - 22 3924 69
XM_017014064.1 XP_016869553.1 21 - 22 3538 69
XM_017014065.1 XP_016869554.1 23 - 24 3658 69
XM_017014066.1 XP_016869555.1 21 - 22 3297 69
XM_017014067.1 XP_016869556.1 21 - 22 3707 69
XM_017014068.1 XP_016869557.1 22 - 23 3498 69
XM_017014069.1 XP_016869558.1 19 - 20 3195 69
XM_017014070.1 XP_016869559.1 18 - 19 3116 69
XM_017014071.1 XP_016869560.1 19 - 20 3364 69
XM_017014072.1 XP_016869561.1 23 - 24 3528 69
XM_017014073.1 XP_016869562.1 23 - 24 3166 69
XM_017014074.1 XP_016869563.1 22 - 23 3449 69
XM_017014075.1 XP_016869564.1 23 - 24 3700 69
XM_017014076.1 XP_016869565.1 23 - 24 3704 69
XM_017014077.1 XP_016869566.1 23 - 24 3341 69
XM_017014078.1 XP_016869567.1 22 - 23 3621 69
XM_017014079.1 XP_016869568.1 22 - 23 4012 69
XM_017014080.1 XP_016869569.1 20 - 21 3253 69
XM_017014081.1 XP_016869570.1 21 - 22 3374 69
XM_017014082.1 XP_016869571.1 21 - 22 3209 69
XM_017014083.1 XP_016869572.1 20 - 21 3049 69
XM_017014084.1 XP_016869573.1 18 - 19 2787 69
GenBank mRNA AB011107.1 - 22 - 23 3350 69
BC117147.1 - 17 - 18 2743 69
BC117149.1 - 17 - 18 2743 69
BC143872.1 - 17 - 18 2738 69
BC143873.1 - 18 - 19 2831 69

View Full Product Details