The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014682.2 | NP_055497.1 | 6 - 7 | 784 | 110 |
GenBank mRNA | AB011107.1 | - | 6 - 7 | 648 | 110 |