The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014700.3 | NP_055515.1 | 2 - 3 | 1142 | 84 |
XM_005255713.3 | XP_005255770.1 | 2 - 3 | 1177 | 84 | |
XM_005255717.3 | XP_005255774.1 | 2 - 3 | 1188 | 84 | |
XM_005255718.3 | XP_005255775.1 | 2 - 3 | 1188 | 84 | |
XM_011522765.2 | XP_011521067.1 | 1 - 2 | 92 | 84 | |
XM_017023907.1 | XP_016879396.1 | 2 - 3 | 1188 | 84 | |
GenBank mRNA | AB014565.1 | - | 2 - 3 | 1190 | 84 |
AF395731.1 | - | 2 - 3 | 1190 | 84 | |
BC051360.1 | - | 2 - 3 | 1141 | 84 |