The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016002.2 | NP_057086.2 | 3 - 4 | 728 | 84 |
GenBank mRNA | AF151807.1 | - | 3 - 4 | 378 | 84 |
AK075178.1 | - | 3 - 4 | 484 | 84 | |
BC026185.2 | - | 3 - 4 | 389 | 84 |