Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLN6
Entrez Gene ID
54982
Gene Name
ceroid-lipofuscinosis, neuronal 6, late infantile, variant
Gene Aliases
CLN4A, HsT18960, nclf
Chromosome Location
Chr.15: 68206992 - 68229742 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017882.2 NP_060352.1 4 - 5 646 91
GenBank mRNA AK000568.1 - 4 - 5 591 91
AK027604.1 - 4 - 5 614 91
AK074753.1 - 1 - 2 140 91
AK291175.1 - 4 - 5 573 91
AK293197.1 - 4 - 5 822 91
BC010849.1 - 4 - 5 560 91
BC013130.1 - 4 - 5 497 91
CR457244.1 - 4 - 5 488 91
KU178671.1 - 4 - 5 488 91
KU178673.1 - 3 - 4 389 91

View Full Product Details