The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018117.11 | NP_060587.8 | 19 - 20 | 2762 | 107 |
XM_005269962.2 | XP_005270019.1 | 19 - 20 | 2479 | 107 | |
XM_005269963.2 | XP_005270020.1 | 19 - 20 | 2611 | 107 | |
XM_005269964.2 | XP_005270021.1 | 12 - 13 | 1517 | 107 | |
XM_017016397.1 | XP_016871886.1 | 18 - 19 | 2450 | 107 | |
XM_017016398.1 | XP_016871887.1 | 18 - 19 | 2380 | 107 | |
XM_017016399.1 | XP_016871888.1 | 18 - 19 | 2324 | 107 | |
GenBank mRNA | AB037772.2 | - | 19 - 20 | 2575 | 107 |
AF320223.1 | - | 19 - 20 | 2549 | 107 | |
AK000579.1 | - | 5 - 6 | 564 | 107 | |
AK001368.1 | - | 14 - 15 | 1665 | 107 | |
AK025050.1 | - | 2 - 3 | 273 | 107 | |
AK299524.1 | - | 12 - 13 | 1382 | 107 | |
AK315145.1 | - | 19 - 20 | 2540 | 107 | |
AL137699.1 | - | 17 - 18 | 2235 | 107 | |
BC040469.1 | - | 19 - 20 | 2579 | 107 | |
BC071564.1 | - | 19 - 20 | 2545 | 107 |