The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039500.2 | NP_001034589.2 | 16 - 17 | 2820 | 90 |
XM_006710360.3 | XP_006710423.2 | 13 - 14 | 2095 | 90 | |
XM_011540683.2 | XP_011538985.1 | 16 - 17 | 2782 | 90 | |
XM_011540684.2 | XP_011538986.1 | 16 - 17 | 2907 | 90 | |
XM_011540685.2 | XP_011538987.1 | 16 - 17 | 2919 | 90 | |
XM_011540688.2 | XP_011538990.1 | 16 - 17 | 2782 | 90 | |
XM_011540690.2 | XP_011538992.1 | 15 - 16 | 2605 | 90 | |
XM_011540691.2 | XP_011538993.1 | 15 - 16 | 2596 | 90 | |
XM_011540692.2 | XP_011538994.1 | 16 - 17 | 2776 | 90 | |
XM_011540693.1 | XP_011538995.1 | 16 - 17 | 2782 | 90 | |
XM_011540695.1 | XP_011538997.1 | 16 - 17 | 2782 | 90 | |
XM_011540697.2 | XP_011538999.1 | 10 - 11 | 2153 | 90 | |
XM_017000305.1 | XP_016855794.1 | 10 - 11 | 1899 | 90 | |
XM_017000306.1 | XP_016855795.1 | 11 - 12 | 2060 | 90 | |
GenBank mRNA | AK057346.1 | - | 5 - 6 | 903 | 90 |
AK295797.1 | - | 16 - 17 | 2804 | 90 | |
BC101378.1 | - | 5 - 6 | 900 | 90 | |
BC101379.2 | - | 5 - 6 | 900 | 90 | |
BC101380.1 | - | 5 - 6 | 900 | 90 | |
BC101381.1 | - | 5 - 6 | 900 | 90 |