The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000059.3 | NP_000050.2 | 5 - 6 | 699 | 142 |
GenBank mRNA | BC026160.1 | - | 5 - 6 | 675 | 142 |
BC047568.1 | - | 5 - 6 | 670 | 142 | |
DQ897648.1 | - | 4 - 5 | 472 | 142 | |
U43746.1 | - | 5 - 6 | 700 | 142 |