The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000093.4 | NP_000084.3 | 25 - 26 | 2673 | 70 |
NM_001278074.1 | NP_001265003.1 | 25 - 26 | 2673 | 70 | |
XM_017014266.1 | XP_016869755.1 | 25 - 26 | 2688 | 70 | |
GenBank mRNA | AB209864.1 | - | 24 - 25 | 2152 | 70 |
AB371583.1 | - | 25 - 26 | 2673 | 70 | |
D90279.1 | - | 25 - 26 | 2416 | 70 |