The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000133.3 | NP_000124.1 | 5 - 6 | 552 | 83 |
NM_001313913.1 | NP_001300842.1 | 4 - 5 | 438 | 83 | |
GenBank mRNA | A13997.1 | - | 5 - 6 | 524 | 83 |
AB186358.1 | - | 4 - 5 | 409 | 83 | |
AK292749.1 | - | 5 - 6 | 523 | 83 | |
BC109214.1 | - | 5 - 6 | 552 | 83 | |
BC109215.1 | - | 5 - 6 | 552 | 83 | |
FR846238.1 | - | 5 - 6 | 523 | 83 | |
FR846239.1 | - | 5 - 6 | 523 | 83 | |
FR846240.1 | - | 5 - 6 | 523 | 83 | |
J00136.1 | - | 5 - 6 | 523 | 83 | |
J00137.1 | - | 5 - 6 | 524 | 83 | |
M11309.1 | - | 5 - 6 | 523 | 83 | |
M35672.1 | - | 4 - 5 | 418 | 83 |