The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000168.5 | NP_000159.3 | 7 - 8 | 1120 | 64 |
XM_011515274.2 | XP_011513576.1 | 6 - 7 | 3744 | 64 | |
XM_017011997.1 | XP_016867486.1 | 6 - 7 | 3777 | 64 | |
GenBank mRNA | AK299299.1 | - | 6 - 7 | 1006 | 64 |
AK308429.1 | - | 8 - 9 | 1185 | 64 | |
BC113616.1 | - | 6 - 7 | 1077 | 64 | |
BC117168.1 | - | 6 - 7 | 1077 | 64 | |
M57609.1 | - | 6 - 7 | 1083 | 64 |