Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLDC
Entrez Gene ID
2731
Gene Name
glycine decarboxylase
Gene Aliases
GCE, GCSP, HYGN1
Chromosome Location
Chr.9: 6532464 - 6645692 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000170.2 NP_000161.2 23 - 24 3029 62
GenBank mRNA AK308470.1 - 9 - 10 1271 62
AK310534.1 - 19 - 20 2404 62
AK314156.1 - 23 - 24 3031 62
BC111993.1 - 23 - 24 2902 62
BC111995.1 - 23 - 24 2902 62
D90239.1 - 23 - 24 2986 62
M63635.1 - 23 - 24 2917 62
M64590.1 - 23 - 24 2986 62

View Full Product Details