Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALB
Entrez Gene ID
213
Gene Name
albumin
Gene Aliases
ANALBA, FDAH, PRO0883, PRO0903, PRO1341
Chromosome Location
Chr.4: 73404255 - 73421412 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000477.5 NP_000468.1 5 - 6 693 94
GenBank mRNA AF190168.1 - 5 - 6 620 94
AK292755.1 - 5 - 6 94
AK298437.1 - 5 - 6 686 94
AK298461.1 - 5 - 6 403 94
AK308044.1 - 5 - 6 535 94
AK314794.1 - 5 - 6 661 94
AY550967.1 - 5 - 6 627 94
AY728024.1 - 5 - 6 626 94
AY960291.1 - 5 - 6 652 94
BC034023.1 - 5 - 6 661 94
BC036003.1 - 5 - 6 661 94
CR749331.1 - 5 - 6 660 94
DQ986150.1 - 5 - 6 631 94
HQ537426.1 - 4 - 5 548 94
V00494.1 - 5 - 6 659 94

View Full Product Details