The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000560.3 | NP_000551.1 | 5 - 6 | 593 | 60 |
NM_001040033.1 | NP_001035122.1 | 6 - 7 | 598 | 60 | |
GenBank mRNA | AK313582.1 | - | 5 - 6 | 546 | 60 |
BC035456.1 | - | 6 - 7 | 580 | 60 | |
BC040693.1 | - | 5 - 6 | 523 | 60 | |
BT019643.1 | - | 4 - 5 | 427 | 60 | |
M37033.1 | - | 5 - 6 | 520 | 60 | |
M60871.1 | - | 5 - 6 | 500 | 60 |