The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000718.3 | NP_000709.1 | 20 - 21 | 3428 | 72 |
NM_001243812.1 | NP_001230741.1 | 20 - 21 | 3428 | 72 | |
XM_011518991.2 | XP_011517293.1 | 20 - 21 | 3395 | 72 | |
XM_011518992.2 | XP_011517294.1 | 20 - 21 | 3392 | 72 | |
XM_011518993.2 | XP_011517295.1 | 20 - 21 | 3391 | 72 | |
XM_011518994.2 | XP_011517296.1 | 20 - 21 | 3394 | 72 | |
XM_017015115.1 | XP_016870604.1 | 20 - 21 | 3392 | 72 | |
XM_017015116.1 | XP_016870605.1 | 20 - 21 | 3395 | 72 | |
XM_017015117.1 | XP_016870606.1 | 20 - 21 | 3394 | 72 | |
XM_017015118.1 | XP_016870607.1 | 20 - 21 | 3388 | 72 | |
XM_017015119.1 | XP_016870608.1 | 20 - 21 | 3394 | 72 | |
XM_017015120.1 | XP_016870609.1 | 20 - 21 | 3391 | 72 | |
GenBank mRNA | AB209467.1 | - | 2 - 3 | 372 | 72 |
M94172.1 | - | 20 - 21 | 3428 | 72 | |
M94173.1 | - | 20 - 21 | 3428 | 72 |