The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000818.2 | NP_000809.1 | 12 - 13 | 1743 | 69 |
NM_001134366.1 | NP_001127838.1 | 12 - 13 | 1743 | 69 | |
GenBank mRNA | AJ251501.1 | - | 9 - 10 | 743 | 69 |
BC126327.1 | - | 12 - 13 | 1265 | 69 | |
BC126329.1 | - | 12 - 13 | 1265 | 69 | |
M74826.1 | - | 12 - 13 | 1381 | 69 | |
M81882.1 | - | 12 - 13 | 1292 | 69 | |
X69936.1 | - | 12 - 13 | 1285 | 69 |