The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000832.6 | NP_000823.4 | 1 - 2 | 591 | 61 |
NM_001185090.1 | NP_001172019.1 | 1 - 2 | 591 | 61 | |
NM_001185091.1 | NP_001172020.1 | 1 - 2 | 591 | 61 | |
NM_007327.3 | NP_015566.1 | 1 - 2 | 591 | 61 | |
NM_021569.3 | NP_067544.1 | 1 - 2 | 591 | 61 | |
XM_005266071.3 | XP_005266128.1 | 1 - 2 | 261 | 61 | |
XM_005266072.3 | XP_005266129.1 | 1 - 2 | 595 | 61 | |
XM_005266073.4 | XP_005266130.1 | 1 - 2 | 656 | 61 | |
XM_011518583.2 | XP_011516885.1 | 1 - 2 | 595 | 61 | |
GenBank mRNA | AB208918.1 | - | 1 - 2 | 338 | 61 |
AB208999.1 | - | 1 - 2 | 573 | 61 | |
AF015730.1 | - | 1 - 2 | 334 | 61 | |
AF015731.1 | - | 1 - 2 | 332 | 61 | |
D13515.1 | - | 1 - 2 | 432 | 61 | |
L05666.1 | - | 1 - 2 | 278 | 61 | |
L13266.1 | - | 1 - 2 | 1354 | 61 | |
L13268.1 | - | 1 - 2 | 231 | 61 |