The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001182.4 | NP_001173.2 | 5 - 6 | 715 | 104 |
NM_001201377.1 | NP_001188306.1 | 5 - 6 | 715 | 104 | |
NM_001202404.1 | NP_001189333.2 | 5 - 6 | 715 | 104 | |
XM_011543417.2 | XP_011541719.1 | 5 - 6 | 706 | 104 | |
XM_017009493.1 | XP_016864982.1 | 5 - 6 | 491 | 104 | |
GenBank mRNA | AK297365.1 | - | 5 - 6 | 661 | 104 |
AK312459.1 | - | 5 - 6 | 544 | 104 | |
BC002515.2 | - | 5 - 6 | 532 | 104 | |
BC071712.1 | - | 5 - 6 | 509 | 104 | |
BC073174.1 | - | 5 - 6 | 501 | 104 | |
S74728.1 | - | 5 - 6 | 493 | 104 |