The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242.4 | NP_001233.1 | 1 - 2 | 346 | 73 |
XM_017020232.1 | XP_016875721.1 | 1 - 2 | 364 | 73 | |
XM_017020234.1 | XP_016875723.1 | 1 - 2 | 364 | 73 | |
GenBank mRNA | AK315732.1 | - | 1 - 2 | 365 | 73 |
BC012160.1 | - | 1 - 2 | 332 | 73 | |
M63928.1 | - | 1 - 2 | 235 | 73 |