The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242.4 | NP_001233.1 | 3 - 4 | 659 | 75 |
XM_011521042.2 | XP_011519344.1 | 2 - 3 | 823 | 75 | |
XM_017020232.1 | XP_016875721.1 | 3 - 4 | 677 | 75 | |
XM_017020233.1 | XP_016875722.1 | 2 - 3 | 835 | 75 | |
XM_017020234.1 | XP_016875723.1 | 3 - 4 | 677 | 75 | |
GenBank mRNA | AK315732.1 | - | 3 - 4 | 678 | 75 |
BC012160.1 | - | 3 - 4 | 645 | 75 | |
M63928.1 | - | 3 - 4 | 548 | 75 |