Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SIGLEC6
Entrez Gene ID
946
Gene Name
sialic acid binding Ig like lectin 6
Gene Aliases
CD327, CD33L, CD33L1, CD33L2, CDW327, OBBP1
Chromosome Location
Chr.19: 51517725 - 51531859 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011527533.2 XP_011525835.1 4 - 5 999 60
XM_011527534.2 XP_011525836.1 4 - 5 1002 60
XM_011527535.2 XP_011525837.1 4 - 5 913 60
XM_011527536.2 XP_011525838.1 4 - 5 838 60
XM_011527537.2 XP_011525839.1 4 - 5 805 60
XM_011527538.2 XP_011525840.1 4 - 5 1001 60
XM_017027511.1 XP_016883000.1 4 - 5 1002 60
GenBank mRNA AK300182.1 - 4 - 5 937 60
AK308174.1 - 4 - 5 913 60
AK309903.1 - 4 - 5 805 60
BC140798.1 - 4 - 5 744 60
D86358.1 - 4 - 5 961 60
D86359.1 - 4 - 5 961 60
U71382.1 - 4 - 5 755 60

View Full Product Details