Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ENTPD6
Entrez Gene ID
955
Gene Name
ectonucleoside triphosphate diphosphohydrolase 6 (putative)
Gene Aliases
CD39L2, IL-6SAG, IL6ST2, NTPDase-6, dJ738P15.3
Chromosome Location
Chr.20: 25195670 - 25228075 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114089.3 NP_001107561.1 2 - 3 523 67
NM_001247.4 NP_001238.2 3 - 4 592 67
NM_001317941.2 NP_001304870.1 3 - 4 589 67
NM_001322378.1 NP_001309307.1 3 - 4 736 67
NM_001322383.1 NP_001309312.1 3 - 4 589 67
NM_001322385.1 NP_001309314.1 2 - 3 523 67
NM_001322386.1 NP_001309315.1 2 - 3 523 67
NM_001322387.1 NP_001309316.1 3 - 4 592 67
NM_001322388.1 NP_001309317.1 3 - 4 592 67
NM_001322389.1 NP_001309318.1 2 - 3 667 67
NM_001322390.1 NP_001309319.1 4 - 5 714 67
NM_001322391.1 NP_001309320.1 3 - 4 592 67
NM_001322392.1 NP_001309321.1 2 - 3 520 67
NM_001322393.1 NP_001309322.1 2 - 3 523 67
NM_001322394.1 NP_001309323.1 2 - 3 520 67
NM_001322395.1 NP_001309324.1 2 - 3 667 67
NM_001322396.1 NP_001309325.1 3 - 4 589 67
NM_001322397.1 NP_001309326.1 3 - 4 592 67
NM_001322398.1 NP_001309327.1 2 - 3 667 67
XM_006723665.2 XP_006723728.1 4 - 5 628 67
XM_011529396.1 XP_011527698.1 4 - 5 632 67
XM_011529397.1 XP_011527699.1 4 - 5 561 67
XM_017028129.1 XP_016883618.1 2 - 3 401 67
XM_017028130.1 XP_016883619.1 2 - 3 401 67
XM_017028131.1 XP_016883620.1 2 - 3 401 67
XM_017028132.1 XP_016883621.1 3 - 4 568 67
XM_017028133.1 XP_016883622.1 2 - 3 371 67
GenBank mRNA AF039916.1 - 3 - 4 526 67
AK074691.1 - 2 - 3 372 67
AK293256.1 - 2 - 3 464 67
AK297954.1 - 3 - 4 432 67
AK302395.1 - 3 - 4 440 67
AK303786.1 - 2 - 3 478 67
AK308626.1 - 2 - 3 490 67
AL834158.1 - 2 - 3 438 67
AY327581.1 - 3 - 4 489 67
BC025980.2 - 3 - 4 496 67
CR936765.1 - 3 - 4 506 67

View Full Product Details