The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001249.3 | NP_001240.1 | 14 - 15 | 1398 | 125 |
NM_001321984.1 | NP_001308913.1 | 15 - 16 | 1525 | 125 | |
NM_001321985.1 | NP_001308914.1 | 15 - 16 | 1515 | 125 | |
NM_001321986.1 | NP_001308915.1 | 15 - 16 | 1495 | 125 | |
NM_001321987.1 | NP_001308916.1 | 16 - 17 | 1642 | 125 | |
NM_001321988.1 | NP_001308917.1 | 14 - 15 | 2174 | 125 | |
XM_005268224.3 | XP_005268281.1 | 14 - 15 | 1360 | 125 | |
XM_006720325.3 | XP_006720388.1 | 14 - 15 | 1401 | 125 | |
XM_017021813.1 | XP_016877302.1 | 15 - 16 | 1520 | 125 | |
XM_017021814.1 | XP_016877303.1 | 12 - 13 | 1275 | 125 | |
XM_017021815.1 | XP_016877304.1 | 15 - 16 | 1520 | 125 | |
GenBank mRNA | AF039918.1 | - | 13 - 14 | 1333 | 125 |
AF136572.1 | - | 13 - 14 | 1369 | 125 | |
AY430094.1 | - | 13 - 14 | 1339 | 125 | |
BC130485.1 | - | 11 - 12 | 1115 | 125 | |
BC130487.1 | - | 11 - 12 | 1115 | 125 |