The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276.2 | NP_001267.2 | 5 - 6 | 634 | 96 |
GenBank mRNA | AK130142.1 | - | 5 - 6 | 544 | 96 |
AK130200.1 | - | 5 - 6 | 544 | 96 | |
AK225266.1 | - | 5 - 6 | 543 | 96 | |
AK225584.1 | - | 5 - 6 | 543 | 96 | |
AK312911.1 | - | 5 - 6 | 617 | 96 | |
BC008568.1 | - | 5 - 6 | 580 | 96 | |
BC038354.1 | - | 5 - 6 | 521 | 96 | |
BC039132.1 | - | 5 - 6 | 530 | 96 | |
BT007223.1 | - | 5 - 6 | 463 | 96 | |
M80927.1 | - | 5 - 6 | 534 | 96 |