Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GCKR
Entrez Gene ID
2646
Gene Name
glucokinase (hexokinase 4) regulator
Gene Aliases
FGQTL5, GKRP
Chromosome Location
Chr.2: 27496839 - 27523689 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001486.3 NP_001477.2 16 - 17 1492 85
XM_017003796.1 XP_016859285.1 11 - 12 969 85
XM_017003797.1 XP_016859286.1 10 - 11 919 85
GenBank mRNA AK291846.1 - 16 - 17 1475 85
AK303784.1 - 15 - 16 1416 85
BC130481.1 - 16 - 17 1472 85
BC130483.1 - 16 - 17 1472 85
Z48475.1 - 16 - 17 1489 85

View Full Product Details