Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GCKR
Entrez Gene ID
2646
Gene Name
glucokinase (hexokinase 4) regulator
Gene Aliases
FGQTL5, GKRP
Chromosome Location
Chr.2: 27496839 - 27523689 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001486.3 NP_001477.2 5 - 6 492 71
XM_011532763.1 XP_011531065.1 5 - 6 492 71
GenBank mRNA AK291846.1 - 5 - 6 481 71
AK309599.1 - 5 - 6 448 71
BC130481.1 - 5 - 6 472 71
BC130483.1 - 5 - 6 472 71
DA011063.1 - 5 - 6 492 71
Z48475.1 - 5 - 6 489 71

View Full Product Details