Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EDNRA
Entrez Gene ID
1909
Gene Name
endothelin receptor type A
Gene Aliases
ET-A, ETA, ETA-R, ETAR, ETRA, MFDA, hET-AR
Chromosome Location
Chr.4: 147480917 - 147544954 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001957.3 NP_001948.1 2 - 3 950 98
NR_045958.1 2 - 3 950 98
GenBank mRNA AK312812.1 - 2 - 3 770 98
AY275462.1 - 1 - 2 420 98
BC022511.1 - 2 - 3 770 98
D90348.1 - 2 - 3 900 98
L06622.1 - 1 - 2 426 98
S45956.1 - 2 - 3 641 98
S57498.1 - 2 - 3 904 98
S63938.1 - 1 - 2 493 98
S67127.1 - 1 - 2 557 98
S81539.1 - 1 - 2 420 98
S81542.1 - 1 - 2 420 98
X61950.1 - 2 - 3 904 98

View Full Product Details