The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002224.3 | NP_002215.2 | 54 - 55 | 7677 | 103 |
XM_011514577.2 | XP_011512879.1 | 53 - 54 | 7317 | 103 | |
GenBank mRNA | AB209739.1 | - | 45 - 46 | 6464 | 103 |
D26351.1 | - | 54 - 55 | 7688 | 103 | |
U01062.1 | - | 54 - 55 | 7489 | 103 |