The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002224.3 | NP_002215.2 | 56 - 57 | 8007 | 93 |
XM_011514577.2 | XP_011512879.1 | 55 - 56 | 7647 | 93 | |
GenBank mRNA | AB209739.1 | - | 47 - 48 | 6794 | 93 |
AK093524.1 | - | 2 - 3 | 888 | 93 | |
D26351.1 | - | 56 - 57 | 8018 | 93 | |
U01062.1 | - | 56 - 57 | 7819 | 93 |