Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OPHN1
Entrez Gene ID
4983
Gene Name
oligophrenin 1
Gene Aliases
ARHGAP41, MRX60, OPN1
Chromosome Location
Chr.X: 68042344 - 68433841 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002547.2 NP_002538.1 12 - 13 1398 99
XM_005262270.1 XP_005262327.1 12 - 13 1468 99
XM_006724653.2 XP_006724716.1 12 - 13 1468 99
XM_011530961.1 XP_011529263.1 12 - 13 1319 99
XM_017029555.1 XP_016885044.1 12 - 13 1468 99
GenBank mRNA AB102656.1 - 11 - 12 1104 99
AJ001189.1 - 12 - 13 1742 99
AK295769.1 - 12 - 13 1468 99
BC140763.1 - 12 - 13 1436 99

View Full Product Details