The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002547.2 | NP_002538.1 | 19 - 20 | 1980 | 94 |
XM_005262270.1 | XP_005262327.1 | 19 - 20 | 2050 | 94 | |
XM_006724653.2 | XP_006724716.1 | 19 - 20 | 2050 | 94 | |
XM_011530961.1 | XP_011529263.1 | 19 - 20 | 1901 | 94 | |
GenBank mRNA | AB102656.1 | - | 18 - 19 | 1686 | 94 |
AJ001189.1 | - | 19 - 20 | 2324 | 94 | |
AK295769.1 | - | 19 - 20 | 2050 | 94 | |
BC140763.1 | - | 19 - 20 | 2018 | 94 |