Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- RANGAP1
- Entrez Gene ID
-
5905
- Gene Name
- Ran GTPase activating protein 1
- Gene Aliases
-
Fug1, RANGAP, SD
- Chromosome Location
-
Chr.22: 41244777 - 41302213 on Build GRCh38
View Full Product Details